Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Epratuzumab Recombinant Human Monoclonal Antibody (hL22 (Epratuzumab))
Human Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA548051
Description
Specificity: This antibody is specific for the 3rd Ig-like domain of human CD22 (epitope B), a cell surface glycoprotein present on mature B-cells and on many types of malignant B-cells.
Epratuzumab is a humanized monoclonal antibody derived from the murine IG2a monoclonal antibody, LL2 (EPB-2). This agent may subsequently be well matched for use in oncology and the treatment of inflammatory autoimmune disorders, such as lupus.
Specifications
Epratuzumab | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
Human | |
Protein A | |
RUO | |
Human, Rhesus Macaque, Cynomolgus Monkey | |
Antibody | |
IgG1 κ |
Flow Cytometry, Inhibition Assays, Immunoprecipitation | |
hL22 (Epratuzumab) | |
Unconjugated | |
90Y-DOTA-hLL2; 90Y-epratuzumab tetraxetan; 90Y-hLL2; AMG-412; hLL2 | |
This antibody was prepared by the humanization of LL2 (EPB-2), a murine anti-CD22 IgG2a raised against Raji Burkitt lymphoma cells. | |
200 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction