Learn More
Invitrogen™ EPS15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579216
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human A375 whole cell, human HepG2 whole cell, human Jurkat whole cell, human PANC-1 whole cell, rat brain tissue, rat liver tissue, mouse brain tissue, mouse spleen tissue, mouse liver tissue, mouse testis tissue.
EPS15 is a ubiquitously expressed protein that is involved in cell growth regulation and may be involved in the regulation of mitogenic signals and control of cell proliferation. Phosphorylation of EPS15 on Tyrosine 849 upon DNA damage is involved in the internalization of EGFR. A chromosomal aberration involving EPS15 is also found in acute leukemias, resulting in a rogue activator protein.
Specifications
EPS15 | |
Polyclonal | |
Unconjugated | |
EPS15 | |
2410112D09Rik; AF1P; AF-1P; AF-1p protein; ALL1 fused gene from chromosome 1; epidermal growth factor pathway substrate 15; epidermal growth factor receptor pathway substrate 15; epidermal growth factor receptor substrate 15; EPS15; MLLT5; protein AF-1p; Protein Eps15; RGD1307641 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13858, 2060, 313474 | |
-20°C | |
Lyophilized |
ELISA, Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and 0.05mg sodium azide | |
P42566, P42567 | |
EPS15 | |
E. coli-derived human EPS15 recombinant protein (Position: A2-E88). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.