Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EPS15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582353
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582353 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582353 Supplier Invitrogen™ Supplier No. PA582353
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CFFRQSTDPF ATSSTDPFSA ANNSSITSVE TLKHNDPFAP GGTVVAASDS ATDPFASVFG NESFGGGFAD FSTLSKVNNE DPFRSATSSS VSNVVITKNV FEETSVKSED EPPALPPKIG TPTRPCPLPP GKRSINKLDS PD Highest antigen sequence indentity to the following orthologs: Mouse - 81%, Rat - 83%.

EPS15 is a ubiquitously expressed protein that is involved in cell growth regulation and may be involved in the regulation of mitogenic signals and control of cell proliferation. Phosphorylation of EPS15 on Tyrosine 849 upon DNA damage is involved in the internalization of EGFR. A chromosomal aberration involving EPS15 is also found in acute leukemias, resulting in a rogue activator protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPS15
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EPS15
Gene Accession No. P42566
Gene Alias 2410112D09Rik; AF1P; AF-1P; AF-1p protein; ALL1 fused gene from chromosome 1; epidermal growth factor pathway substrate 15; epidermal growth factor receptor pathway substrate 15; epidermal growth factor receptor substrate 15; EPS15; MLLT5; protein AF-1p; Protein Eps15; RGD1307641
Gene Symbols EPS15
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EPS15. Recombinant protein control fragment (Product #RP-89543).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2060
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.