Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPS15 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981208
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1208 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1208 Supplier Bioss Supplier No. BS11251R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EPS15 is a ubiquitously expressed protein that is involved in cell growth regulation and may be involved in the regulation of mitogenic signals and control of cell proliferation. Phosphorylation of EPS15 on Tyrosine 849 upon DNA damage is involved in the internalization of EGFR. A chromosomal aberration involving EPS15 is also found in acute leukemias, resulting in a rogue activator protein.

Specifications

Antigen EPS15
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene EPS15
Gene Accession No. P42566
Gene Alias 2410112D09Rik; AF1P; AF-1P; AF-1p protein; ALL1 fused gene from chromosome 1; epidermal growth factor pathway substrate 15; epidermal growth factor receptor pathway substrate 15; Epidermal growth factor receptor substrate 15; Eps15; MLLT5; Protein AF-1p; Protein Eps15; RGD1307641
Gene Symbols EPS15
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human EPS15.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2060
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.