Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPS15R Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10088149
Description
EPS15R Polyclonal specifically detects EPS15R in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
EPS15R | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
epidermal growth factor receptor pathway substrate 15-like 1, epidermal growth factor receptor substrate 15-like 1, epidermal growth factor receptor substrate EPS15R, EPS15R, Eps15-related protein | |
Rabbit | |
Affinity Purified | |
RUO | |
58513 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 2 - 5 ug/mg lysate | |
NP_067058.1 | |
EPS15L1 | |
The immunogen recognized by this antibody maps to a region between residue 814 and 864 of human epidermal growth factor receptor pathway substrate 15-like 1 using the numbering given in entry NP_067058.1 (GeneID 58513). | |
0.1 mL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction