Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EPS15R Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553965
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553965 Supplier Invitrogen™ Supplier No. PA553965
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KQDAQDRLDE MDQQKAKLRD MLSDVRQKCQ DETQMISSLK TQIQSQESDL KSQEDDLNRA KSELNRLQQE ETQLEQSIQA GRVQLETIIK SLKSTQDEIN QARSKLSQ Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

Seems to be a constitutive component of clathrin-coated pits that is required for receptor-mediated endocytosis. Involved in endocytosis of integrin beta-1 (ITGB1) and transferrin receptor (TFR); internalization of ITGB1 as DAB2-dependent cargo but not TFR seems to require association with DAB2. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPS15R
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EPS15L1
Gene Accession No. Q9UBC2
Gene Alias 9830147J04Rik; AI593686; epidermal growth factor receptor pathway substrate 15 like 1; epidermal growth factor receptor pathway substrate 15-like 1; epidermal growth factor receptor pathway substrate 15-related sequence; Epidermal growth factor receptor substrate 15-like 1; epidermal growth factor receptor substrate EPS15R; Eps15l1; EPS15R; eps15-related protein; Eps15-rs
Gene Symbols EPS15L1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EPS15R. Recombinant protein control fragment (Product #RP-92469).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58513
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.