Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPS15R Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109690
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109690 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109690 Supplier Thermo Scientific Supplier No. PA5109690
Add to Cart
Add to Cart

EPS15R Polyclonal Antibody

EPS15R Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen EPS15R
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EPS15L1
Gene Accession No. Q9UBC2
Gene Alias 9830147J04Rik; AI593686; epidermal growth factor receptor pathway substrate 15 like 1; epidermal growth factor receptor pathway substrate 15-like 1; epidermal growth factor receptor pathway substrate 15-related sequence; Epidermal growth factor receptor substrate 15-like 1; epidermal growth factor receptor substrate EPS15R; Eps15l1; EPS15R; eps15-related protein; Eps15-rs
Gene Symbols EPS15L1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 475-754 of human EPS15L1 (NP_001245304.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58513
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.