Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eps8 Antibody (18F5B11) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280724
Description
Eps8 Monoclonal specifically detects Eps8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Eps8 | |
Monoclonal | |
0.5 mg/mL | |
Western Blot : 1-5 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin : 5 μg/mL | |
epidermal growth factor receptor kinase substrate 8, epidermal growth factor receptor pathway substrate 8 | |
Mouse | |
Protein G purified | |
RUO | |
2059 | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
18F5B11 | |
Unconjugated | |
PBS | |
EPS8 | |
Amino acids 701-882 of human EPS8 protein were used as the immunogen for the EPS8 antibody. | |
0.1 mg | |
Primary | |
Human | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction