Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EPS8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582121
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582121 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582121 Supplier Invitrogen™ Supplier No. PA582121
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PKEQFIPPYV PRFRNGWEPP MLNFMGATME QDLYQLAESV ANVAEHQRKQ EIKRLSTEHS SVSEYHPADG YAFSSNIYTR GSHLDQGEAA VAFKPTSNRH IDRNYEPLKT QPKKYAKSKY DFVARNNSEL SVLKDDILEI LDD Highest antigen sequence indentity to the following orthologs: Rat - 80%, Mouse - 80%.

This gene encodes a member of the EPS8 family. This protein contains one PH domain and one SH3 domain. It functions as part of the EGFR pathway, though its exact role has not been determined. Highly similar proteins in other organisms are involved in the transduction of signals from Ras to Rac and growth factor-mediated actin remodeling. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPS8
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EPS8
Gene Accession No. Q12929
Gene Alias AI042819; AW261790; AW545405; DFNB102; epidermal growth factor receptor kinase substrate 8; Epidermal growth factor receptor kinase substrate 8-like protein 2; epidermal growth factor receptor pathway substrate 8; epidermal growth factor receptor pathway substrate 8-like protein 2; epidermal growth factor receptor pathway substrate 8-related protein 2; EPS8; EPS8 related protein 2; Eps8l2; EPS8-like 2; EPS8-like protein 2; Eps8r2; EPS8-related protein 2
Gene Symbols EPS8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EPS8. Recombinant protein control fragment (Product #RP-102267).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2059
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.