Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPS8L3 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298054V
Description
EPS8L3 Polyclonal specifically detects EPS8L3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
EPS8L3 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
epidermal growth factor receptor kinase substrate 8-like protein 3, Epidermal growth factor receptor pathway substrate 8-related protein 3, EPS8-like 3, EPS8-like protein 3, EPS8R3, EPS8-related protein 3, FLJ21522, MGC16817 | |
Produced in rabbits immunized with E. coli-derived Human EPS8L3 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
79574 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction