Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EPS8L3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556611
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556611 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556611 Supplier Invitrogen™ Supplier No. PA556611
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WLVKNEAGRS GYIPSNILEP LQPGTPGTQG QSPSRVPMLR LSSRPEEVTD WLQAENFSTA TVRTLGSLTG SQLLRIRPGE LQMLCPQE Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.

This gene encodes a protein that is related to epidermal growth factor receptor pathway substrate 8 (EPS8), a substrate for the epidermal growth factor receptor. The function of this protein is unknown. Alternatively spliced transcript variants encoding different isoforms exist. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPS8L3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EPS8L3
Gene Accession No. Q8TE67
Gene Alias AI504489; Epidermal growth factor receptor kinase substrate 8-like protein 3; epidermal growth factor receptor pathway substrate 8 related protein 3; epidermal growth factor receptor pathway substrate 8-related protein 3; EPS8 like 3; Eps8l3; EPS8-like 3; EPS8-like protein 3; Eps8r3; EPS8-related protein 3; ESP8-like 3
Gene Symbols EPS8L3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EPS8L3. Recombinant protein control fragment (Product #RP-94553).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79574
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.