Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Epsin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119098
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119098 200 μL
PIPA5122012 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119098 Supplier Invitrogen™ Supplier No. PA5119098
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Epsin 1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

This gene encodes a member of the epsin protein family. The encoded protein binds clathrin and is involved in the endocytosis of clathrin-coated vesicles. Loss of function of this gene is associated with reduced tumor growth and progression in certain cancer types.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Epsin 1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene EPN1
Gene Accession No. Q80VP1, Q9Y6I3
Gene Alias AA269831; EH domain-binding mitotic phosphoprotein; Epn1; EPS-15-interacting protein 1; Epsin 1; epsin-1; hypothetical protein LOC508199; Ibp1; Intersectin-EH-binding protein 1; LOW QUALITY PROTEIN: epsin-1
Gene Symbols EPN1
Host Species Rabbit
Immunogen Recombinant protein Epsin 1. The antigen corresponds to amino acid range 279-437 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13854, 29924
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.