Learn More
Epsin 1 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304525A

Description
The recommended shelf life for this product is 1 year from date of receipt.
EPN1 is an endocytic accessory protein that interacts with EPS15 (MIM 600051), the alpha subunit of the clathrin adaptor AP2 (AP2A1; MIM 601026), and clathrin (see MIM 118960), as well as with other accessory proteins for the endocytosis of clathrin-coated vesicles.Specifications
Epsin 1 | |
Polyclonal | |
Unconjugated | |
EPN1 | |
EH domain-binding mitotic phosphoprotein, EPS-15-interacting protein 1, epsin-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13854, 29924 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q80VP1, Q9Y6I3 | |
EPN1 | |
Between 475 and 525 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.