Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epsin-2 Antibody (OTI1G3), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271701MFV610
Description
Epsin-2 Monoclonal antibody specifically detects Epsin-2 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Flow CytometrySpecifications
Epsin-2 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Primate, Monkey | |
Purified |
Flow Cytometry | |
OTI1G3 | |
Flow Cytometry | |
Eps15 binding protein, EPS-15-interacting protein 2, epsin 2, epsin-2, KIAA1065EHB21 | |
Full length human recombinant protein of human EPN2(NP_055779) produced in HEK293T cell. | |
0.1 mL | |
Cell Biology, Signal Transduction | |
22905 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction