Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epsin 2 Polyclonal Antibody
Epsin 2 Polyclonal Antibody
Supplier: Thermo Scientific PA531158
Description
Epsin 2 Polyclonal Antibody for Western Blot

Specifications
Epsin 2 | |
Polyclonal | |
Unconjugated | |
EPN2 | |
9530051D10Rik; AA536924; EH domain binding protein epsin 2; EHB21; EPN2; Eps15 binding protein; EPS-15-interacting protein 2; epsin 2; epsin-2; Ibp2; Intersectin-EH-binding protein 2; KIAA1065 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22905 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.69 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O95208 | |
EPN2 | |
Recombinant protein fragment corresponding to a region within amino acids 55 and 265 of Human Epsin 2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction