Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Epsin 3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580593
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human EPN3.
EPN3 belongs to the epsin family.
Specifications
Epsin 3 | |
Polyclonal | |
Unconjugated | |
EPN3 | |
2310022G12Rik; Epn3; EPS-15-interacting protein 3; epsin 3; epsin-3 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
55040 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with no preservative | |
Q9H201 | |
EPN3 | |
A synthetic peptide corresponding to the center region of the Human EPN3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction