Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epstein Barr Virus Antibody (CS1) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308575
Description
Epstein Barr Virus Monoclonal specifically detects Epstein Barr Virus in Virus samples. It is validated for Western Blot, ELISA.Specifications
Epstein Barr Virus | |
Monoclonal | |
1 mg/mL | |
Western Blot : 1-2 ug/mL, ELISA | |
Epstein Barr Virus, Human herpesvirus type 4, Hypothetical protein BALF1 | |
Recombinant fusion protein containing the sequence of bacterial beta-galactosidase and the carboxyl half of EBV-encoded LMP | |
100 μg | |
Primary | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, ELISA | |
CS1 | |
Unconjugated | |
10mM PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction