Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epstein Barr Virus Antibody (CS2), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308759UV
Description
Epstein Barr Virus Monoclonal antibody specifically detects Epstein Barr Virus in Virus samples. It is validated for Western Blot, ELISASpecifications
Epstein Barr Virus | |
Monoclonal | |
DyLight 350 | |
Epstein Barr Virus, Human herpesvirus type 4, Hypothetical protein BALF1 | |
Recombinant fusion protein containing the sequence of bacterial beta-galactosidase and the carboxyl half of EBV-encoded LMP | |
100 μg | |
Immune System Diseases, Immunology, Virology Bacteria and Parasites | |
Virus | |
Purified |
Western Blot, ELISA | |
CS2 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction