Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Epstein Barr Virus BGLF4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567990
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567990 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567990 Supplier Invitrogen™ Supplier No. PA567990
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Human samples.

BGLF4 plays many key roles by phosphorylating several proteins including the viral DNA processivity factor BMRF1, EBNA1 or EBNA2. It modifies the host nuclear envelope structure and induces the redistribution of nuclear envelope-associated proteins by phosphorylating host nucleoporins. Subsequently, promotes the nuclear transport of EBV lytic proteins. Required for efficient lytic DNA replication and release of nucleocapsids from the nucleus. Contributes to the compaction of host cell chromatin in cells undergoing lytic replication, presumably by phosphorylating the host condensin complex and host TOP2A. Induces disassembly of the nuclear lamina by phosphorylating with host LMNA. Phosphorylates substrates involved in capsid assembly and DNA packaging. Facilitates the switch from latent to lytic DNA replication by down-regulating EBNA1 replication function. Phosphorylates the viral immediate-early protein BZLF1 and inhibits its sumoylation by interacting with host SUMO1 and SUMO2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Epstein Barr Virus BGLF4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Alias EBV
Host Species Rabbit
Immunogen Synthesized peptide derived from human KR2_EBV.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.