Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Epstein Barr Virus BHRF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117549
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117549 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117549 Supplier Invitrogen™ Supplier No. PA5117549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

Prevents premature death of the host cell during virus production, which would otherwise reduce the amount of progeny virus. Acts as a host B-cell leukemia/lymphoma 2 (Bcl-2) homolog, and interacts with pro-apoptotic proteins to prevent mitochondria permeabilization, release of cytochrome c and subsequent apoptosis of the host cell.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Epstein Barr Virus BHRF1
Applications Western Blot
Classification Polyclonal
Concentration 2.535 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene BHRF1
Gene Accession No. P03182
Gene Alias Apoptosis regulator BHRF1; EA-R; Early antigen protein R; Nuclear antigen
Gene Symbols BHRF1
Host Species Rabbit
Immunogen Recombinant Epstein-Barr virus Apoptosis regulator BHRF1 protein (1-142aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3783706
Target Species Virus
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.