Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Epstein Barr Virus BLLF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117556
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117556 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117556 Supplier Invitrogen™ Supplier No. PA5117556
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

Involved in nucleotide metabolism: produces dUMP, the immediate precursor of thymidine nucleotides and decreases the intracellular concentration of dUTP to avoid uracil incorporation into viral DNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Epstein Barr Virus BLLF3
Applications Western Blot
Classification Polyclonal
Concentration 3.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Alias BLLF3; Deoxyuridine 5'-triphosphate nucleotidohydrolase; dUTP pyrophosphatase
Host Species Rabbit
Immunogen Recombinant Epstein-Barr virus Deoxyuridine 5'-triphosphate nucleotidohydrolase protein (1-278aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.