Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Epstein Barr Virus BZLF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117635
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117635 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117635 Supplier Invitrogen™ Supplier No. PA5117635
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

Plays a role in virion attachment to host B-lymphocytes, through binding to leukocyte antigen (HLA) class II and subsequently participates in fusion of the virion with host membranes. May act as a tropism switch that directs fusion with B-lymphocytes and inhibits fusion with epithelial cells. Additionally, hampers T-cell recognition via HLA class II molecules through steric hindrance of T-cell receptor-class II-peptide interaction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Epstein Barr Virus BZLF2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene BZLF2
Gene Accession No. P03205
Gene Alias Glycoprotein 42; gp42
Gene Symbols BZLF2
Host Species Rabbit
Immunogen Recombinant Epstein-Barr virus Glycoprotein 42 protein (34-223aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3783745
Target Species Virus
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.