Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Epstein Barr Virus glycoprotein B Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144512
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144512 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144512 Supplier Invitrogen™ Supplier No. PA5144512
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Epstein Barr Virus glycoprotein B Polyclonal Antibody for Western Blot, ELISA

Envelope glycoprotein that forms spikes at the surface of virion envelope. Essential for the initial attachment to heparan sulfate moieties of the host cell surface proteoglycans. Involved in fusion of viral and cellular membranes leading to virus entry into the host cell. Following initial binding to its host receptors, membrane fusion is mediated by the fusion machinery composed at least of gB and the heterodimer gH/gL. May be involved in the fusion between the virion envelope and the outer nuclear membrane during virion egress.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Epstein Barr Virus glycoprotein B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.468 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene gB
Gene Accession No. P03188
Gene Alias Envelope glycoprotein B; gB
Gene Symbols gB
Host Species Rabbit
Immunogen Recombinant Epstein-Barr virus gB protein (23-260aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3783680
Target Species Virus
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.