Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epstein-Barr virus gp350 Mouse anti-Epstein-Barr virus, Clone: EA070, Abnova™

Catalog No. 89033506 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-033-506 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-033-506 Supplier Abnova Corporation Supplier No. MAB13543
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against Epstein-Barr virus (EBV) gp350.

  • High specificity
  • High reactivity

Specifications

Antigen Epstein-Barr virus gp350
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone EA070
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Epstein-Barr virus (EBV) gp350.
Dilution ELISA Immunofluorescence The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4.
Host Species Mouse
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with gp350 of Epstein-Barr virus.
Target Species Epstein-Barr virus (EBV)
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.