Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epstein Barr Virus Mouse anti-Virus, Clone: CS1, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30779220UG
Description
Epstein Barr Virus Monoclonal specifically detects Epstein Barr Virus in Virus samples. It is validated for Western Blot, ELISA.Specifications
Epstein Barr Virus | |
Monoclonal | |
0.2 mg/ml | |
Western Blot 1-2 ug/ml, ELISA | |
Epstein Barr Virus, Human herpesvirus type 4, Hypothetical protein BALF1 | |
Recombinant fusion protein containing the sequence of bacterial beta-galactosidase and the carboxyl half of EBV-encoded LMP | |
20 μg | |
Primary | |
Store at 4°C. | |
IgG1 κ |
Western Blot, ELISA | |
CS1 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction