Learn More
Invitrogen™ Epstein Barr Virus Nuclear Antigen 1 Polyclonal Antibody, Biotin

Description
The PA1-7499 antibody reacts with Epstein-Barr virus EBNA-1. PA1-7499 has been successfully used in Immunohistochemical, ELISA, Immunofluorescence and Western blot applications. The PA1-7499 antibody was raised against Recombinant EBV EBNA-1. Store at 4°C for up to 6 months. For long term storage, aliquot and freeze at -20°C. Avoid freeze-thaw cycles.
Specifications
Specifications
| Antigen | Epstein Barr Virus Nuclear Antigen 1 |
| Applications | ELISA |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | EBNA-1 |
| Host Species | Goat |
| Immunogen | Purified recombinant EBV EBNA-1. |
| Purification Method | IgG fraction |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.