Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Epstein Barr Virus Nuclear Antigen 1 Polyclonal Antibody, HRP
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA17498
Description
The PA1-7498 antibody reacts with Epstein-Barr virus EBNA-1. PA1-7498 has been successfully used in ELISA applications. The PA1-7498 antibody was raised against Recombinant EBV EBNA-1. Note: Sodium azide is a potent inhibitor of peroxidase.
Epstein-Barr virus (EBV) is a human herpes virus, which is associated with conditions such as Hodgkin's disease and Burkitt's Lymphoma and is the causative agent in mononucleosis in adolescents. EBV latently infects B lymphocytes. Infected B cells express EBV nuclear antigens and latent proteins LMP1, LMP2A and LMP2B. LMP2A forms aggregates in the plasma membranes of B lymphocytes, where it functions as a negative regulator of the Src and Syk protein tyrosine kinases. Studies show that LMP2A blocks B-cell receptor (BCR) signal transduction in EBV immortalized B cells in vitro and may play an important role in maintaining a latent EBV infection within the peripheral blood B cells of infected individuals.EBNA-1 is the only viral protein consistently expressed in all malignancies associated with EBV.
Specifications
Epstein Barr Virus Nuclear Antigen 1 | |
Polyclonal | |
HRP | |
EBNA-1 | |
Purified recombinant EBV EBNA-1. | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 10mg/mL BSA and 0.002% thimerosal | |
Goat | |
IgG fraction | |
RUO | |
Virus | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction