Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Epstein Barr Virus p18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA173003
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA173003 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA173003 Supplier Invitrogen™ Supplier No. PA173003
Add to Cart
Add to Cart

Goat Polyclonal Antibody

The PA1-73003 antibody reacts with Epstein-Barr virus p18. PA1-73003 has been successfully used in ELISA and immunofluorescence applications. IHC and WB are potential applications but have not yet been tested. The PA1-73003 antibody was raised against Recombinant EBV p18. Recommended short term storage (<6 months) is at 4°C. For longer term storage, aliquot and freeze.

Epstein-Barr virus (EBV) is a human herpes virus, which is associated with conditions such as Hodgkin's disease and Burkitt's Lymphoma and is the causative agent in mononucleosis in adolescents. EBV latently infects B lymphocytes. Infected B cells express EBV nuclear antigens and latent proteins LMP1, LMP2A and LMP2B. LMP2A forms aggregates in the plasma membranes of B lymphocytes, where it functions as a negative regulator of the Src and Syk protein tyrosine kinases. Studies show that LMP2A blocks B-cell receptor (BCR) signal transduction in EBV immortalized B cells in vitro and may play an important role in maintaining a latent EBV infection within the peripheral blood B cells of infected individuals.EBNA-1 is the only viral protein consistently expressed in all malignancies associated with EBV.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Epstein Barr Virus p18
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 5.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias EBV p18
Host Species Goat
Immunogen Recombinant EBV p18.
Purification Method IgG fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.