Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPX Antibody (EPX/3908R), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308476MFV610
Description
EPX Monoclonal antibody specifically detects EPX in Human samples. It is validated for Flow CytometrySpecifications
EPX | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
8288 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
EPX/3908R | |
Flow Cytometry | |
EC 1.11.1.7, eosinophil peroxidase, EPOEPER, EPPEC 1.11.1, EPX-PEN | |
Human eosinophils from a patient with hypereosinophilic syndrome (Uniprot: P11678 ) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction