Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EPX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562200
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562200 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562200 Supplier Invitrogen™ Supplier No. PA562200
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EGTDPASPGA VETSVLRDCI AEAKLLVDAA YNWTQKS Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 78%.

EPX belongs to the peroxidase family, XPO subfamily. Defects in EPX are the cause of eosinophil peroxidase deficiency (EPD).
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPX
Applications Western Blot
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EPX
Gene Accession No. P11678
Gene Alias eosinophil peroxidase; Eosinophil peroxidase heavy chain; Eosinophil peroxidase light chain; EPER; EPO; EPP; Epx; EPX PEN; EPXD; EPX-PEN; PERE
Gene Symbols EPX
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EPX. Recombinant protein control fragment (Product #RP-101234).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8288
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.