Learn More
Invitrogen™ EQTN Polyclonal Antibody

Description
Immunogen sequence: EDVNKQEEKN EDHTPNYAPA NEKNGNYYKD IKQYVFTTQN PNGTESEISV RATTDLNFAL KNGSTPNVPA FWTMLAKAIN GTAVVMDDKD QLFHPIPESD VNATQGENQP DLEDLKIK Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 50%.
Specifications
Specifications
| Antigen | EQTN |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | EQTN |
| Gene Accession No. | Q9NQ60 |
| Gene Alias | acrosome formation associated factor; acrosome formation-associated factor; AFAF; C9orf11; EQTN; Equatorin; equatorin, sperm acrosome associated; SPACA8; sperm acrosome associated 8 |
| Gene Symbols | EQTN |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.