Learn More
Cytiva Equilibration Tubes
Catalog No. 45002610 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
12/Pk.
Catalog No. Quantity
45-002-610 12/Pk.
1 options
Catalog No. 45-002-610 Supplier Cytiva Supplier No. 80646779
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
Includes
Recommendation regarding equilibration solution volumes
Provides a convenient means for equilibrating IPG strips prior to 2-dimension electrophoresis
  • Complete packsize: 12
TRUSTED_SUSTAINABILITY
Specifications
For Use With For equilibrating IPG strips prior to 2-dimension electrophoresis
Quantity 12/Pk.
Product Type Equilibration Tube