Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Equine Donor Serum

Catalog No. p-2625236
Click to view available options
Quantity:
500 mL
Color:
Golden Brown

Specifications

Certifications/Compliance U.S. regulations
Color Golden Brown
Content And Storage -20°C
Species Horse
Form Liquid
Product Type Horse Serum
Quantity 500 mL
Sterility Sterile
Sterilization Method 0.2 μm filtration
For Use With (Application) Diagnostic assays
Hemoglobin Concentration ≤35mg/dL
Mycoplasma Detected No
Osmolality 273 to 301 mOsm/kg
pH 7.1 to 8
Protein Content 5.6 to 9.1 g/dL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.