Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Equine IL-1 beta/IL-1F2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB33401
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB33401 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB33401 Supplier R&D Systems Supplier No. MAB33401
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-1 beta/IL-1F2 Monoclonal specifically detects IL-1 beta/IL-1F2 in Equine samples. It is validated for Immunocytochemistry.

Specifications

Antigen IL-1 beta/IL-1F2
Applications Immunocytochemistry
Classification Monoclonal
Clone 608714
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunocytochemistry 20-30 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q28286
Gene Alias catabolin, IL-1, IL1 beta, IL-1 beta, IL-1b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Gene Symbols IL1B
Host Species Mouse
Immunogen E. coli-derived recombinant equine IL-1 beta/IL-1F2 Ala116-Ala268 Accession # Q28286
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3553
Test Specificity Detects equine IL-1 beta/IL-1F2 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant canine, guinea pig, mouse, rat, and porcine IL-1 beta is observed and 10-50% cross-reactivity with recombinant feline, human, rabbit, and rhesus IL-1 beta is observed. Approximately 20% cross-reactivity with recombinant mouse (rm) IL-36 alpha and no cross-reactivity with recombinant human (rh) IL-36 alpha, rhIL-36 gamma, rhIL-1F10, rhIL-36Ra, rmIL-1Ra, rmIL-18, or rmIL-36 beta is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Equine
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.