Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Equine IL-10 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1605SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1605SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1605SP Supplier R&D Systems Supplier No. AF1605SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IL-10 Polyclonal specifically detects IL-10 in Equine samples. It is validated for Western Blot, Neutralization, Immunocytochemistry, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen IL-10
Applications Western Blot, Neutralization, Immunocytochemistry, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 0.2-0.6 ug/mL, Immunocytochemistry 10-25 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q28374
Gene Alias CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF
Gene Symbols IL10
Host Species Goat
Immunogen E. coli-derived recombinant equine IL-10 Ser19-Asn178 Accession # Q28374
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3586
Test Specificity Detects equine IL-10 in ELISAs and Western blots. In sandwich immunoassays, less than 1% cross-reactivity with recombinant canine IL-10 and recombinant porcine IL-10 is observed and less than 0.4% cross-reactivity with recombinant human IL-10, recombinant mouse IL-10, recombinant rat IL-10, and recombinant feline IL-10 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Equine
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.