Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Equine IL-1ra/IL-1F3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2466SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2466SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2466SP Supplier R&D Systems Supplier No. AF2466SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IL-1ra/IL-1F3/IL1RN Polyclonal specifically detects IL-1ra/IL-1F3/IL1RN in Equine samples. It is validated for Western Blot, Immunocytochemistry, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen IL-1ra/IL-1F3/IL1RN
Applications Western Blot, Immunocytochemistry, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O18999
Gene Alias DIRA, ICIL-1RA, IL1F3, IL-1F3, IL-1ra, IL1RA, IL-1ra3, IL1RN, IL-1RN, interleukin 1 receptor antagonist, IRAP, MVCD4
Gene Symbols IL1RN
Host Species Goat
Immunogen E. coli-derived recombinant equine IL-1ra/IL-1F3 His26-Gln177 Accession # O18999
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3557
Test Specificity Detects equine IL-1ra/IL-1F3 in ELISAs and Western blots. In sandwich immunoassays, less than 20% cross-reactivity with recombinant mouse IL-1ra is observed, less than 5% cross-reactivity with recombinant rat IL-1ra is observed, less than 2% cross-reactivity with recombinant porcine IL-1ra is observed, less than 0.4% cross-reactivity with recombinant human IL-1ra is observed, and less than 0.2% cross-reactivity with recombinant equine IL-1beta is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Equine
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.