Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Equine IL-1ra/IL-1F3 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF2466
This item is not returnable.
View return policy
Description
IL-1ra/IL-1F3/IL1RN Polyclonal specifically detects IL-1ra/IL-1F3/IL1RN in Equine samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
IL-1ra/IL-1F3/IL1RN | |
Polyclonal | |
Biotin | |
O18999.1 | |
IL1RN | |
Recombinant Equine IL-1ra/IL-1F3 His26-Gln177 Accession # O18999.1 | |
50 μg | |
Primary | |
Detects equine IL-1ra in ELISAs and Western blots. In sandwich ELISAs, approximately 20% cross-reactivity is observed with rmIL-1ra, less than 5% cross-reactivity with rrIL-1ra, less than 2% cross-reactivity with rpIL-1ra, and less than 0.4% cross-reactivity with rhIL-1ra is observed. | |
Equine | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
DIRA, ICIL-1RA, IL1F3, IL-1F3, IL-1ra, IL1RA, IL-1ra3, IL1RN, IL-1RN, interleukin 1 receptor antagonist, IRAP, MVCD4 | |
Goat | |
Affinity Purified | |
RUO | |
3557 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction