Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Equine IL-2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF1613
This item is not returnable.
View return policy
Description
IL-2 Polyclonal specifically detects IL-2 in Equine samples. It is validated for Western Blot, Immunocytochemistry, ELISA Capture (Matched Antibody Pair).Specifications
IL-2 | |
Polyclonal | |
Unconjugated | |
NP_001078902.1 | |
IL2 | |
E. coli-derived recombinant equine IL-2 Ala21-Thr149 (Cys141Ser) Accession # NP_001078902.1 | |
100 μg | |
Primary | |
Detects equine IL-2 in ELISAs and Western blots. In sandwich immunoassays, less than 0.3% cross-reactivity with recombinant human IL-2, recombinant mouse IL-2, recombinant rat IL-2, recombinant feline IL-2, recombinant bovine IL-2, recombinant canine IL-2, recombinant cotton rat IL-2, and recombinant porcine IL-2 is observed. | |
Equine | |
IgG |
Western Blot, Immunocytochemistry, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
aldesleukin, IL2, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, Proleukin, T cell growth factor, T-cell growth factor, TCGF | |
Goat | |
Affinity Purified | |
RUO | |
3558 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction