Learn More
Invitrogen™ Equine IL-4 Recombinant Protein
Description
Mature equine IL-4 was expressed in Pichia pastoris, X-33. GenBank™ Accession # P42202, amino acids 24 - 132. For protein verification, the expressed protein was N-terminal sequenced and begins with the following 10 amino acids: CKYDITLQEI. Activity: The biological activity of recombinant equine IL-4 was determined by measuring the proliferation of TF-1 cells. The ED50 (or 1 unit of activity) for this effect in our laboratories typically falls into the range of 20 - 50 ng/mL. Ref: Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. Uses: For in vitro applications. A dose-response assay should be performed to determine the optimal concentration for specific experiments. Reconstitute with 0.5 mL of distilled water. Reconstitute with 0.5 mL of distilled or deionized water. For dilution, use a buffer containing carrier protein, e.g. PBS with 1% BSA or a neutral media containing 10% FCS and freeze in aliquots at -70°C.
Specifications
Specifications
| Accession Number | P42202 |
| For Use With (Application) | ELISA, Immunoprecipitation, Western Blot |
| Formulation | PBS with no preservative |
| Gene ID (Entrez) | 100034225 |
| Molecular Weight (g/mol) | 12.3 kDa |
| Name | Equine IL-4 |
| Quantity | 20 μg |
| Source | Yeast |
| Regulatory Status | RUO |
| Gene Alias | B cell growth factor 1; B_cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor; B-cell stimulatory factor 1; BCGF1; BCGF-1; binetrakin; BSF1; BSF-1; cytokine; H-IL-4; IGG1 induction factor; IL2; IL4; Il-4; Il4e12; ILN; Interleukin; interleukin 4; interleukin 4 variant 2; interleukin 4 variant IL-4 delta2; interleukin 4 variant IL-4 delta3; interleukin 4 variant IL-4 int2A; interleukin 4 variant IL-4 int3A; interleukin 4 variant IL-4 int3B; Interleukin4; interleukin-4; interleukin-4 precursor; lymphocyte stimulatory factor 1; MGC79402; M-IL-4; Pitrakinra |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.