Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Equine Progesterone ELISA Kit (Colorimetric)
SDP

Catalog No. NB601191KT Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB601191KT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB601191KT Supplier Novus Biologicals™ Supplier No. NBP2601191KIT
Add to Cart
Add to Cart

High sensitivity and specificity for detection of Equine horse progesterone.

The Equine Progesterone ELISA Kit (Colorimetric) measures equine Progesterone in serum, plasma.

Specifications

Assay Range 0.25 to 100ng/mL
Assay Sensitivity 0.25ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Plasma, Serum
Specificity For detection of horse PROG
Target Species Equine
For Use With (Application) ELISA
Kit Contents Instruction manual, Standard, Stop Solution, Wash Buffer (20 x concentrate), Substrate B, Substrate A, HRP-conjugate, Assay plate, Antibody, Adhesive Strip (For 96 wells)
Quantity 1 Kit
Regulatory Status RUO
Research Discipline Signal Transduction
Sample Volume 50 to 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Synonym Pregn 4 ene 3 20 dione
Target Progesterone
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.