Learn More
Description
Specifications
Specifications
| Assay Range | 1.875 to 30ng/mL |
| Assay Sensitivity | 1.402ng/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Plasma, Serum |
| Target Species | Equine |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Regulatory Status | RUO |
| Research Discipline | Lipid and Metabolism |
| Sample Volume | 50 to 100 μL |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
