Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ER 27319 maleate
GREENER_CHOICE

Catalog No. 247110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-711-0 10 mg
24-715-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 24-711-0 Supplier Tocris Bioscience™ Supplier No. 2471/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective Syk kinase inhibitor

Selective inhibitor of Syk kinase. Inhibits tyrosine phosphorylation of Syk initiated by the engagement of FcεRI in rat and human mast cells. Results in the abrogation of degranulation, TNF-α production (IC50 = 10 μM) and other related signaling events.

Chemical Identifiers

CAS 1204480-26-1
Molecular Formula C22H24N2O5

Specifications

Quantity 10 mg
Formula Weight 396.17
Purity >99%
Molecular Weight (g/mol) 396.443
Chemical Name or Material ER 27319 maleate
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.