Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ER alpha/NR3A1 Antibody (ER505), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP233106AF700
Description
ER alpha/NR3A1 Monoclonal specifically detects ER alpha/NR3A1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
ER alpha/NR3A1 | |
Monoclonal | |
Alexa Fluor 700 | |
ER, ER alpha, Era, ER-alpha, ESRESRA, Estradiol receptor, estrogen receptor, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 3*4,56,7*/819-2 isoform, estrogen receptor alpha delta 4 +49 isoform, estrogen receptor alpha delta 4*5,6,7*/654 isoform, NR3A1DKFZp686N23123, Nuclear receptor subfamily 3 group A member 1 | |
Mouse | |
67 kDa | |
0.1 mL | |
Angiogenesis, Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
2099 | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
ER505 | |
Western Blot, ELISA, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
ESR1 | |
Recombinant human ER alpha/NR3A1 alpha protein (aa2-185) (Uniprot: P03372) | |
Protein A or G purified | |
RUO | |
Primary | |
This monoclonal antibody is specific to ER alpha and shows minimal cross-reaction with other members of the family. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy. This monoclonal antibody is excellent for staining of formalin-fixed, paraffin-embedded breast carcinomas. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction