Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ER alpha/NR3A1 Antibody (ER505), Novus Biologicals™

Mouse Monoclonal Antibody
$487.50
Specifications
Antigen | ER alpha/NR3A1 |
---|---|
Clone | ER505 |
Concentration | 0.2mg/mL |
Dilution | Western Blot 0.5-1ug/ml, Immunohistochemistry-Paraffin 1-2ug/ml, SDS-Page |
Applications | Western Blot, Immunohistochemistry (Paraffin), SDS-Page |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP23273301
![]() |
Novus Biologicals
NBP2327330.1MG |
0.1 mg |
Each for $487.50
|
|
|||||
NBP23273302
![]() |
Novus Biologicals
NBP2327330.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
ER alpha/NR3A1 Monoclonal specifically detects ER alpha/NR3A1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
ER alpha/NR3A1 | |
0.2mg/mL | |
Western Blot, Immunohistochemistry (Paraffin), SDS-Page | |
Unconjugated | |
Mouse | |
Human | |
ER, ER alpha, Era, ER-alpha, ESRESRA, Estradiol receptor, estrogen receptor, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 3*4,56,7*/819-2 isoform, estrogen receptor alpha delta 4 +49 isoform, estrogen receptor alpha delta 4*5,6,7*/654 isoform, NR3A1DKFZp686N23123, Nuclear receptor subfamily 3 group A member 1 | |
ESR1 | |
IgG1 κ | |
Protein G purified | |
This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy. This MAb is excellent for staining of formalin-fixed, paraffin-embedded breast carcinomas. |
ER505 | |
Western Blot 0.5-1ug/ml, Immunohistochemistry-Paraffin 1-2ug/ml, SDS-Page | |
Monoclonal | |
Purified | |
RUO | |
PBS with 0.05% BSA. with 0.05% Sodium Azide | |
2099 | |
Recombinant human Estrogen Receptor alpha protein (aa2-185) | |
Primary | |
Store at 4C. | |
67 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title