Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ER alpha/NR3A1 Antibody (h-151), Novus Biologicals™

Mouse Monoclonal Antibody
$181.90 - $487.50
Specifications
| Antigen | ER alpha/NR3A1 |
|---|---|
| Clone | h-151 |
| Concentration | 1.0 mg/mL |
| Applications | Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018070
![]() |
Novus Biologicals
NB120135380.025MG |
0.025 mg |
Each for $181.90
|
|
|||||
NB12013538
![]() |
Novus Biologicals
NB12013538 |
0.1 mg |
Each for $487.50
|
|
|||||
Description
ER alpha/NR3A1 Monoclonal specifically detects ER alpha/NR3A1 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| ER alpha/NR3A1 | |
| 1.0 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| ER, ER alpha, Era, ER-alpha, ESRESRA, Estradiol receptor, estrogen receptor, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 3*4,56,7*/819-2 isoform, estrogen receptor alpha delta 4 +49 isoform, estrogen receptor alpha delta 4*5,6,7*/654 isoform, NR3A1DKFZp686N23123, Nuclear receptor subfamily 3 group A member 1 | |
| ESR1 | |
| IgG1 | |
| Protein G purified |
| h-151 | |
| Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Signal Transduction, Transcription Factors and Regulators | |
| P03372 | |
| 2099 | |
| Synthetic peptide derived from the sequence of human Estrogen Receptor, conjugated to KLH | |
| Primary | |
| Binding specificity was confirmed in peptide inhibition studies with the intact protein. This does not recognize the native estrogen receptor in the inactive 8S oligomeric state, but reacts efficiently with the activated 4S receptor. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title