Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ER beta/NR3A2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300747
Description
ER beta/NR3A2 Polyclonal specifically detects ER beta/NR3A2 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunofluorescence.Specifications
ER beta/NR3A2 | |
Polyclonal | |
Unconjugated | |
PBS, 1mg/ml BSA with 0.05% Sodium Azide | |
Erb, ER-beta, ESR-BETA, ESTRB, estrogen receptor 2 (ER beta), estrogen receptor beta, estrogen receptor beta 4, NR3A2ESRB, Nuclear receptor subfamily 3 group A member 2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Detects estrogen receptor (ER) beta from human and rat tissues. Detects a band at ∼ 55 kDa. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunofluorescence | |
1 mg/mL | |
Western Blot 1 ug/ml, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 2 - 5 ug/ml, Immunohistochemistry-Frozen 2 - 5 ug/ml, Immunofluorescence 1:100 | |
Q92731 | |
ESR2 | |
Synthetic peptide corresponding to residues C(459) S P A E D S K S K E G S Q N P Q S Q(477) of human ER beta. | |
50 μg | |
Cancer, GPCR | |
2100 | |
Human, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction