Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ER beta/NR3A2 Antibody (ESR2/7006R), Janelia Fluor™ 549, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP314205JF549
This item is not returnable.
View return policy
Description
ER beta/NR3A2 Monoclonal antibody specifically detects ER beta/NR3A2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
ER beta/NR3A2 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
ESR2/7006R | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Erb, ER-beta, ESR-BETA, ESTRB, estrogen receptor 2 (ER beta), estrogen receptor beta, estrogen receptor beta 4, NR3A2ESRB, Nuclear receptor subfamily 3 group A member 2 | |
C-terminus fragment of recombinant human ER beta/NR3A2 protein (Uniprot: Q92731 ) | |
0.1 mL | |
Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Signal Transduction, Transcription Factors and Regulators | |
2100 | |
Store at 4°C in the dark. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction