Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ER beta/NR3A2 Antibody (PPG5/10), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261944JF646
Description
ER beta/NR3A2 Monoclonal specifically detects ER beta/NR3A2 in Human, Rat, Porcine, Avian, Primate, Cynomolgus Monkey, Reptile, Rhesus Macaque, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| ERbeta/NR3A2 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ESR2 | |
| Synthetic peptide CEDSKSKEGSQNPQSQ derived from the C terminus of human ER beta1 | |
| 0.1 mL | |
| Primary | |
| Human, Rat, Pig, Avian, Primate, Cynomolgus Monkey, Reptile, Rhesus Macaque, Sheep | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| PPG5/10 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Erb, ER-beta, ESR-BETA, ESTRB, estrogen receptor 2 (ER beta), estrogen receptor beta, estrogen receptor beta 4, NR3A2ESRB, Nuclear receptor subfamily 3 group A member 2 | |
| Mouse | |
| Protein A purified | |
| Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Signal Transduction, Transcription Factors and Regulators | |
| 2100 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction