Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ER beta/NR3A2 Antibody (PPG5/25) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | ER beta/NR3A2 |
---|---|
Clone | PPG5/25 |
Concentration | 1 mg/mL |
Dilution | Western Blot, Immunohistochemistry 1:50 - 1:100, Immunocytochemistry/Immunofluorescence 1:500 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
Description
ER beta/NR3A2 Monoclonal specifically detects ER beta/NR3A2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
ER beta/NR3A2 | |
1 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
PBS with 0.02% Sodium Azide | |
2100 | |
Tuberculin conjugated peptide that recognizes C terminus of ER beta5 at sequence LLSHVRHARYAP | |
Primary |
PPG5/25 | |
Western Blot, Immunohistochemistry 1:50 - 1:100, Immunocytochemistry/Immunofluorescence 1:500 | |
Monoclonal | |
Purified | |
Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Signal Transduction, Transcription Factors and Regulators | |
Erb, ER-beta, ESR-BETA, ESTRB, estrogen receptor 2 (ER beta), estrogen receptor beta, estrogen receptor beta 4, NR3A2ESRB, Nuclear receptor subfamily 3 group A member 2 | |
ESR2 | |
IgG1 | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title