Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ER-HR3 Antibody (ER-HR3), DyLight 550

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB10064931R
Description
ER-HR3 Monoclonal specifically detects ER-HR3 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
| ER-HR3 | |
| Monoclonal | |
| DyLight 550 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Rat | |
| 0.125 mL | |
| NB100-64931 recognizes the murine antigen ER-HR3. ER-HR3 is a cell surface antigen which is expressed by Langerhans cells in epithelium, a subset of mature macrophages and dendritic cells that are located predominantly in haematopoietic and lymphoid organs. Initial studies of ER-HR3 demonstrated expression on peripheral blood monocytes but recent studies suggest that expression is actually very low. During foetal development, ER-HR3 positive cells are localised to haemapoietic islands and are often associated with erythroid progenitor cells. The functions of the ER-HR3 antigen have not been established but reports suggest that the antigen may be involved in adult erythropoiesis and in the regulation of the immune response. Clone ER-HR3 does not inhibit T cell proliferation in antigen-specific T-cell proliferation studies. Clone ER-HR3 recognizes two proteins of approximately 69kD and 55kD under non-reducing conditions. | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| ER-HR3 | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
| Hematopoiesis related macrophage | |
| Adherent F1 (CBAxBL) bone marrow stormal cells. | |
| Primary | |
| Mouse | |
| IgG2c |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction